Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPS18B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014046.3 | NP_054765.1 | 2 | 282 | ||
GenBank mRNA | AF100761.1 | - | 2 | 137 | ||
AF151017.1 | - | 2 | 163 | |||
AK294086.1 | - | 2 | 144 | |||
AK311890.1 | - | 2 | 133 | |||
AL050361.1 | - | 2 | 141 | |||
BC005373.1 | - | 2 | 162 | 3933607 | ||
BG773408.1 | - | 2 | 273 | |||
BP369345.1 | - | 2 | 282 |