Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OGN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014057.4 | NP_054776.1 | 2 | 362 | ||
NM_024416.4 | NP_077727.3 | 2 | 281 | |||
NM_033014.3 | NP_148935.1 | 2 | 644 | |||
GenBank mRNA | AF086912.1 | - | 2 | 209 | ||
AF100758.1 | - | 2 | 209 | |||
AF202167.1 | - | 1 | 134 | |||
AK075521.1 | - | 1 | 173 | |||
AK129845.1 | - | 1 | 166 | |||
AK289628.1 | - | 2 | 359 | |||
AK295433.1 | - | 2 | 281 | |||
AL110267.1 | - | 1 | 151 | |||
BC037273.2 | - | 1 | 150 | 5265880 | ||
BC095443.1 | - | 1 | 166 | 30405707 | ||
BG772245.1 | - | 2 | 283 | |||
BX646984.1 | - | 2 | 319 | |||
CR533516.1 | - | 1 | 83 |