Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HCK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172129.1 | NP_001165600.1 | 7 | 818 | ||
NM_001172130.1 | NP_001165601.1 | 7 | 815 | |||
NM_001172131.1 | NP_001165602.1 | 7 | 815 | |||
NM_001172132.1 | NP_001165603.1 | 8 | 884 | |||
NM_001172133.1 | NP_001165604.1 | 8 | 882 | |||
NM_002110.3 | NP_002101.2 | 7 | 818 | |||
GenBank mRNA | AK026432.1 | - | 7 | 752 | ||
AK225819.1 | - | 7 | 764 | |||
AK289896.1 | - | 7 | 806 | |||
AK290928.1 | - | 8 | 830 | |||
AK298726.1 | - | 8 | 828 | |||
AK314335.1 | - | 7 | 731 | |||
BC014435.1 | - | 7 | 704 | 4855747 | ||
BC094847.1 | - | 7 | 715 | 5923744 | ||
BC108930.1 | - | 7 | 745 | 40025497 | ||
BC108931.1 | - | 7 | 745 | 40025500 | ||
BC113854.1 | - | 7 | 742 | 40025503 | ||
BC114463.1 | - | 7 | 745 | 40025502 | ||
M16592.1 | - | 6 | 593 |