Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FSHR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000145.3 | NP_000136.2 | 1, 2 | 255 | ||
NM_181446.2 | NP_852111.2 | 1, 2 | 255 | |||
XM_011532733.2 | XP_011531035.1 | 1, 2 | 243 | |||
XM_011532734.2 | XP_011531036.1 | 1, 2 | 255 | |||
XM_011532740.1 | XP_011531042.1 | 1, 2 | 243 | |||
GenBank mRNA | AK292562.1 | - | 1, 2 | 223 | ||
AK310367.1 | - | 1, 2 | 255 | |||
AY429104.1 | - | 1, 2 | 145 | |||
BC096831.1 | - | 1, 2 | 145 | 7001973 | ||
BC118548.1 | - | 1, 2 | 178 | 40007014 | ||
BC125270.1 | - | 1, 2 | 178 | 40007010 | ||
DB064473.1 | - | 1, 2 | 255 | |||
JN003607.1 | - | 1, 2 | 182 | |||
M65085.1 | - | 1, 2 | 211 | |||
M95489.1 | - | 1, 2 | 220 | |||
S59900.1 | - | 1, 2 | 219 | |||
X68044.1 | - | 1, 2 | 161 |