Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMARCAL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127207.1 | NP_001120679.1 | 3 | 306 | ||
NM_014140.3 | NP_054859.2 | 3 | 492 | |||
XM_005246631.2 | XP_005246688.1 | 3 | 430 | |||
XM_005246632.1 | XP_005246689.1 | 3 | 482 | |||
XM_017004228.1 | XP_016859717.1 | 3 | 291 | |||
GenBank mRNA | AF082179.1 | - | 2 | 262 | ||
AF432223.1 | - | 3 | 302 | |||
AK023780.1 | - | 3 | 433 | |||
BC016482.1 | - | 2 | 253 | 4341673 | ||
BC043341.1 | - | 3 | 279 | 6058575 | ||
DA856812.1 | - | 3 | 306 | |||
DB099423.1 | - | 3 | 492 |