Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUDT5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321647.1 | NP_001308576.1 | 3 | 500 | ||
NM_001321648.1 | NP_001308577.1 | 3 | 500 | |||
NM_014142.3 | NP_054861.2 | 3 | 500 | |||
GenBank mRNA | AF155832.1 | - | 3 | 246 | ||
AF161464.1 | - | 3 | 248 | |||
AF218818.1 | - | 3 | 203 | |||
AK291131.1 | - | 3 | 236 | |||
AK314144.1 | - | 3 | 486 | |||
BC000025.2 | - | 3 | 411 | 3505623 | ||
BU602000.1 | - | 3 | 502 | |||
CR457195.1 | - | 2 | 84 |