Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LAT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014146.3 | NP_054865.2 | 2, 3 | 463 | ||
NM_032463.2 | NP_115852.1 | 3, 4 | 582 | |||
NM_032464.2 | NP_115853.2 | 3, 4 | 652 | |||
XM_011516558.2 | XP_011514860.1 | 3, 4 | 492 | |||
GenBank mRNA | AF161531.1 | - | 2, 3 | 215 | ||
AF252611.1 | - | 3, 4 | 403 | |||
AF252612.1 | - | 2, 3 | 214 | |||
AF252613.1 | - | 2, 3 | 199 | |||
AF252614.1 | - | 3, 4 | 333 | |||
AF257135.1 | - | 1, 2 | 134 | |||
AK002099.1 | - | 3, 4 | 442 | |||
AK092904.1 | - | 3, 4 | 793 | |||
AK225150.1 | - | 2, 3 | 214 | |||
AK290074.1 | - | 3, 4 | 330 | |||
AK290916.1 | - | 2, 3 | 211 | |||
AY190023.1 | - | 1, 2 | 144 | |||
BC001609.1 | - | 3, 4 | 464 | 3543310 | ||
BC009204.2 | - | 1, 2 | 136 | 4054137 |