Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC113 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142302.1 | NP_001135774.1 | 1, 2 | 180 | ||
NM_014157.3 | NP_054876.2 | 1, 2 | 180 | |||
XM_011523042.2 | XP_011521344.1 | 1, 2 | 180 | |||
GenBank mRNA | AF161550.1 | - | 1, 2 | 139 | ||
AK299066.1 | - | 1, 2 | 180 | |||
AK314300.1 | - | 1, 2 | 169 | |||
AL136785.1 | - | 1, 2 | 157 | |||
BC126445.1 | - | 1, 2 | 149 | 8992161 | ||
BC126447.1 | - | 1, 2 | 149 | 8992163 | ||
CR533506.1 | - | 1, 2 | 101 | |||
DC369542.1 | - | 1, 2 | 181 |