Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPL18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318817.1 | NP_001305746.1 | 3 | 404 | ||
NM_014161.4 | NP_054880.2 | 3 | 404 | |||
NR_134860.1 | 4 | 524 | ||||
NR_134861.1 | 3 | 481 | ||||
NR_134862.1 | 3 | 404 | ||||
NR_134863.1 | 3 | 388 | ||||
NR_134864.1 | 3 | 287 | ||||
NR_134865.1 | 3 | 404 | ||||
NR_134866.1 | 3 | 404 | ||||
GenBank mRNA | AF161556.1 | - | 3 | 382 | ||
AK311391.1 | - | 4 | 401 | |||
AL136633.1 | - | 3 | 354 | |||
AV756740.1 | - | 3 | 339 | |||
BC001623.1 | - | 3 | 375 | 3357333 | ||
BF797811.1 | - | 3 | 287 | |||
BG548185.1 | - | 3 | 383 | |||
BM924328.1 | - | 3 | 497 | |||
BU601065.1 | - | 3 | 345 | |||
BU932963.1 | - | 3 | 393 | |||
BU943173.1 | - | 407 | ||||
CA777760.1 | - | 3 | 286 | |||
CD675657.1 | - | 3 | 287 | |||
CR533491.1 | - | 3 | 262 |