Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ROM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000327.3 | NP_000318.1 | 3 | 1470 | ||
GenBank mRNA | AK313674.1 | - | 3 | 1470 | ||
BC008100.1 | - | 3 | 1060 | 4177529 | ||
BC036268.1 | - | 3 | 1060 | |||
L07894.1 | - | 3 | 934 |