Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC6A16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014037.2 | NP_054756.2 | 2 | 347 | ||
XM_005258820.2 | XP_005258877.1 | 3 | 448 | |||
XM_006723168.3 | XP_006723231.1 | 2 | 363 | |||
XM_011526859.2 | XP_011525161.2 | 2 | 381 | |||
XM_011526860.2 | XP_011525162.1 | 3 | 660 | |||
XM_017026712.1 | XP_016882201.1 | 2 | 363 | |||
GenBank mRNA | AF151977.1 | - | 1 | 152 | ||
AF265578.1 | - | 1 | 113 | |||
AL136856.1 | - | 1 | 159 | |||
BC034948.1 | - | 2 | 315 | 4821848 | ||
BG772801.1 | - | 2 | 354 |