Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPS18B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014046.3 | NP_054765.1 | 2 | 321 | ||
GenBank mRNA | AF100761.1 | - | 2 | 176 | ||
AF151017.1 | - | 2 | 202 | |||
AK294086.1 | - | 2 | 183 | |||
AK311890.1 | - | 2 | 172 | |||
AL050361.1 | - | 2 | 180 | |||
BC005373.1 | - | 2 | 201 | 3933607 | ||
BG773408.1 | - | 2 | 312 | |||
BP369345.1 | - | 2 | 321 |