Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPL42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014050.3 | NP_054769.1 | 5 | 483 | ||
NM_172177.3 | NP_751917.1 | 5 | 486 | |||
NR_038159.1 | 6 | 506 | ||||
NR_038160.1 | 6 | 503 | ||||
NR_038161.1 | 6 | 588 | ||||
GenBank mRNA | AF078860.1 | - | 5 | 465 | ||
AF135160.1 | - | 5 | 469 | |||
AF151038.1 | - | 5 | 457 | |||
AK000285.1 | - | 5 | 459 | |||
AL136659.1 | - | 5 | 460 | |||
BC040240.2 | - | 4 | 422 | 3445213 | ||
BM147739.1 | - | 5 | 484 | |||
BM726733.1 | - | 6 | 461 | |||
BU561400.1 | - | 482 | ||||
CD385500.1 | - | 5 | 479 | |||
CR533495.1 | - | 4 | 324 | |||
CR749344.1 | - | 6 | 560 |