Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NCOA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242539.2 | NP_001229468.1 | 3, 4 | 571 | ||
NM_001318240.1 | NP_001305169.1 | 6, 7 | 3093 | |||
NM_014071.4 | NP_054790.2 | 5, 6 | 863 | |||
XM_006723755.3 | XP_006723818.1 | 5, 6 | 791 | |||
XM_017027742.1 | XP_016883231.1 | 5, 6 | 793 | |||
XM_017027744.1 | XP_016883233.1 | 5, 6 | 790 | |||
XM_017027751.1 | XP_016883240.1 | 5, 6 | 791 | |||
GenBank mRNA | AF128458.1 | - | 4, 5 | 724 | ||
AF171667.1 | - | 4, 5 | 666 | |||
AF177388.1 | - | 4, 5 | 682 | |||
AF208227.1 | - | 6, 7 | 3090 | |||
AF245115.1 | - | 3, 4 | 581 | |||
BC136272.1 | - | 4, 5 | 672 | 9020259 | ||
D80003.2 | - | 4, 5 | 659 | |||
JF707637.1 | - | 3, 4 | 522 | |||
JF707638.1 | - | 3, 4 | 522 | |||
JF707639.1 | - | 3, 4 | 522 |