Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNTNAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014141.5 | NP_054860.1 | 3 | 828 | ||
XM_017011950.1 | XP_016867439.1 | 3 | 1154 | |||
GenBank mRNA | AB020675.1 | - | 3 | 337 | ||
AF319045.1 | - | 3 | 452 | |||
AK315113.1 | - | 3 | 417 | |||
BC093780.1 | - | 3 | 355 | 7939625 | ||
BC113373.1 | - | 3 | 355 | 8322425 | ||
KJ534811.1 | - | 2 | 201 | |||
KJ535069.1 | - | 312 |