Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZC3H7A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014153.3 | NP_054872.2 | 3, 4 | 273 | ||
XM_006720877.2 | XP_006720940.1 | 3, 4 | 309 | |||
XM_011522463.1 | XP_011520765.1 | 4, 5 | 458 | |||
GenBank mRNA | AK000325.1 | - | 1, 2 | 13 | ||
AK091803.1 | - | 3, 4 | 245 | |||
AY163807.1 | - | 2, 3 | 302 | |||
BC036857.1 | - | 3, 4 | 217 | |||
BC046363.1 | - | 3, 4 | 166 | 5538820 | ||
DA160892.1 | - | 3, 4 | 273 |