Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPS18B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014046.3 | NP_054765.1 | 7 | 1362 | ||
GenBank mRNA | AF100761.1 | - | 7 | 1217 | ||
AF151017.1 | - | 7 | 1247 | |||
AK294086.1 | - | 6 | 1157 | |||
AL050361.1 | - | 5 | 1442 |