Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACAD9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014049.4 | NP_054768.2 | 10 | 1196 | ||
NR_033426.1 | 10 | 1372 | ||||
XM_017006264.1 | XP_016861753.1 | 10 | 1360 | |||
XM_017006265.1 | XP_016861754.1 | 4 | 346 | |||
GenBank mRNA | AB209427.1 | - | 10 | 2893 | ||
AF327351.1 | - | 10 | 1026 | |||
AK022568.1 | - | 9 | 1122 | |||
AK024012.1 | - | 10 | 1040 | |||
AK130311.1 | - | 10 | 1067 | |||
AK294597.1 | - | 10 | 864 | |||
AK297952.1 | - | 10 | 1240 | |||
AK310644.1 | - | 9 | 969 | |||
BC001817.2 | - | 3 | 160 | |||
BC007970.2 | - | 10 | 1073 | 4304209 | ||
BC013354.2 | - | 10 | 1050 | 3935925 |