Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NTMT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286796.1 | NP_001273725.1 | 3 | 692 | ||
NM_001286797.1 | NP_001273726.1 | 3 | 505 | |||
NM_001286798.1 | NP_001273727.1 | 3 | 566 | |||
NM_001286799.1 | NP_001273728.1 | 3 | 495 | |||
NM_001286802.1 | NP_001273731.1 | 2 | 350 | |||
NM_001286803.1 | NP_001273732.1 | 2 | 324 | |||
NM_014064.3 | NP_054783.2 | 3 | 540 | |||
XM_005251939.3 | XP_005251996.1 | 3 | 481 | |||
XM_017014642.1 | XP_016870131.1 | 3 | 398 | |||
GenBank mRNA | AF110776.1 | - | 3 | 483 | ||
AK290957.1 | - | 3 | 537 | |||
AK292332.1 | - | 3 | 542 | |||
AK313829.1 | - | 3 | 530 | |||
BC001396.2 | - | 3 | 519 | 3050940 | ||
BC033234.1 | - | 3 | 494 | 5426447 | ||
BI083998.1 | - | 3 | 505 | |||
BM009113.1 | - | 2 | 277 | |||
BM554138.1 | - | 3 | 495 | |||
CN406947.1 | - | 3 | 553 | |||
HY058177.1 | - | 2 | 346 |