Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSORS1C1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014068.2 | NP_054787.2 | 6 | 542 | ||
GenBank mRNA | AB031479.1 | - | 6 | 527 | ||
AF484418.1 | - | 6 | 543 | |||
AF484419.1 | - | 2 | 148 | |||
BC104870.1 | - | 4 | 284 | 8143873 | ||
BI825917.1 | - | 3 | 204 | |||
DB454051.1 | - | 4 | 329 |