Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD274 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267706.1 | NP_001254635.1 | 4 | 487 | ||
NM_014143.3 | NP_054862.1 | 5 | 829 | |||
GenBank mRNA | AF177937.1 | - | 4 | 721 | ||
AF233516.1 | - | 5 | 773 | |||
AK001894.1 | - | 3 | 536 | |||
AK300470.1 | - | 6 | 684 | |||
AK314567.1 | - | 5 | 790 | |||
AY254342.1 | - | 4 | 721 | |||
AY291313.1 | - | 3 | 379 | |||
BC069381.1 | - | 5 | 771 | 7262208 | ||
BC074984.2 | - | 4 | 741 | 30915301 | ||
BC113734.1 | - | 4 | 742 | 8322786 | ||
BC113736.1 | - | 4 | 742 | 8322788 | ||
DQ286582.1 | - | 3 | 379 |