Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZC3H7A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014153.3 | NP_054872.2 | 5 | 570 | ||
XM_006720877.2 | XP_006720940.1 | 5 | 606 | |||
XM_011522463.1 | XP_011520765.1 | 6 | 755 | |||
XM_017023174.1 | XP_016878663.1 | 5 | 587 | |||
XM_017023175.1 | XP_016878664.1 | 2 | 265 | |||
GenBank mRNA | AK000325.1 | - | 3 | 310 | ||
AK091803.1 | - | 5 | 542 | |||
AY163807.1 | - | 4 | 599 | |||
BC036857.1 | - | 5 | 514 | |||
BC046363.1 | - | 5 | 463 | 5538820 |