Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SETD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014159.6 | NP_054878.5 | 11, 12 | 5452 | ||
XM_011533631.2 | XP_011531933.1 | 12, 13 | 6948 | |||
XM_011533632.2 | XP_011531934.1 | 11, 12 | 5524 | |||
XM_011533633.2 | XP_011531935.1 | 11, 12 | 6815 | |||
XM_011533634.1 | XP_011531936.1 | 11, 12 | 5477 | |||
XM_017006270.1 | XP_016861759.1 | 9, 10 | 6651 | |||
GenBank mRNA | AB051519.2 | - | 9, 10 | 3452 | ||
AF161554.1 | - | 9, 10 | 1196 | |||
AJ238403.1 | - | 9, 10 | 4299 | |||
AL831959.1 | - | 8, 9 | 4427 | |||
AY576987.1 | - | 9, 10 | 3961 | |||
AY576988.1 | - | 10, 11 | 3964 | |||
BC090954.1 | - | 9, 10 | 3242 | |||
BC117162.1 | - | 9, 10 | 4075 | 40125713 | ||
BC117164.1 | - | 9, 10 | 4075 | 40125715 | ||
BX649110.1 | - | 9, 10 | 5219 |