Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPL18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318817.1 | NP_001305746.1 | 4 | 795 | ||
NM_014161.4 | NP_054880.2 | 4 | 944 | |||
NR_134860.1 | 5 | 1064 | ||||
NR_134861.1 | 4 | 1021 | ||||
NR_134862.1 | 4 | 944 | ||||
NR_134863.1 | 4 | 928 | ||||
NR_134864.1 | 4 | 827 | ||||
NR_134865.1 | 4 | 795 | ||||
NR_134866.1 | 4 | 773 | ||||
GenBank mRNA | AF161556.1 | - | 4 | 922 | ||
AL136633.1 | - | 4 | 894 | |||
BC001623.1 | - | 4 | 915 | 3357333 |