Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPL22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014990.2 | NP_001014990.1 | 4 | 196 | ||
NM_014180.3 | NP_054899.2 | 5 | 314 | |||
GenBank mRNA | AF161507.1 | - | 5 | 282 | ||
AK000601.1 | - | 5 | 301 | |||
AK097880.1 | - | 1 | 1951 | |||
AK302784.1 | - | 4 | 176 | |||
BC012565.1 | - | 5 | 284 | 4333117 | ||
BP372420.1 | - | 5 | 314 | |||
BU943770.1 | - | 5 | 307 | |||
CR933679.1 | - | 4 | 175 |