Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPR18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098200.1 | NP_001091670.1 | 2 | 1012 | ||
NM_005292.3 | NP_005283.1 | 3 | 1204 | |||
XM_006719946.3 | XP_006720009.1 | 3 | 2014 | |||
GenBank mRNA | AF261135.1 | - | 2 | 887 | ||
AF339781.1 | - | 2 | 960 | |||
BC008569.1 | - | 2 | 971 | 4338810 | ||
BC050646.1 | - | 3 | 1150 | 4341573 | ||
BC066927.1 | - | 3 | 1260 | 4373724 | ||
BT009907.1 | - | 1 | 703 | |||
JQ308176.1 | - | 1 | 703 |