Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC6A7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014228.4 | NP_055043.2 | 6 | 1282 | ||
XM_017009767.1 | XP_016865256.1 | 7 | 1189 | |||
XM_017009768.1 | XP_016865257.1 | 7 | 1265 | |||
XM_017009769.1 | XP_016865258.1 | 7 | 2796 | |||
XM_017009770.1 | XP_016865259.1 | 7 | 916 | |||
GenBank mRNA | AK096607.1 | - | 6 | 1186 | ||
AK308895.1 | - | 6 | 1058 | |||
AK312309.1 | - | 6 | 1062 | |||
BC069631.1 | - | 6 | 815 | 7262437 | ||
BC093785.1 | - | 6 | 891 | 7939630 | ||
BC113425.1 | - | 6 | 891 | 8322477 | ||
S80071.1 | - | 6 | 815 |