Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAPN6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014289.3 | NP_055104.2 | 3 | 478 | ||
GenBank mRNA | AF029232.1 | - | 3 | 378 | ||
AJ000388.1 | - | 3 | 469 | |||
AK300340.1 | - | 2 | 227 | |||
AK312701.1 | - | 3 | 442 | |||
BC000730.2 | - | 2 | 307 | 3503916 | ||
DA863483.1 | - | 3 | 478 |