Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ORC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014321.3 | NP_055136.1 | 1, 2 | 133 | ||
XM_011522978.2 | XP_011521280.1 | 1, 2 | 123 | |||
GenBank mRNA | AF139658.1 | - | 1, 2 | 121 | ||
AK021946.1 | - | 1, 2 | 94 | |||
AK024019.1 | - | 1, 2 | 127 | |||
AK024077.1 | - | 1, 2 | 120 | |||
BC039032.1 | - | 1, 2 | 113 | 5577396 | ||
BC063565.1 | - | 1, 2 | 102 | 3460593 |