Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AMACR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167595.1 | NP_001161067.1 | 2 | 406 | ||
NM_014324.5 | NP_055139.4 | 2 | 406 | |||
NM_203382.2 | NP_976316.1 | 2 | 406 | |||
GenBank mRNA | AF047020.1 | - | 2 | 398 | ||
AF158378.1 | - | 2 | 375 | |||
AJ130733.1 | - | 2 | 396 | |||
AK000912.1 | - | 2 | 391 | |||
AK022765.1 | - | 2 | 376 | |||
AK222831.1 | - | 2 | 378 | |||
AK225749.1 | - | 2 | 415 | |||
AK292988.1 | - | 2 | 356 | |||
AL545355.3 | - | 2 | 319 | |||
AY330488.1 | - | 2 | 319 | |||
AY330489.1 | - | 2 | 319 | |||
AY935981.1 | - | 2 | 319 | |||
BC009471.1 | - | 2 | 322 | 2958112 | ||
BG708726.1 | - | 2 | 342 | |||
BI550368.1 | - | 2 | 444 | |||
BT007193.1 | - | 2 | 310 | |||
DA156454.1 | - | 2 | 409 | |||
EF560721.1 | - | 2 | 329 | |||
FJ498906.1 | - | 2 | 322 | |||
FJ498907.1 | - | 2 | 322 | |||
FJ498908.1 | - | 2 | 322 |
Gene Symbol: C1QTNF3-AMACR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037951.1 | 7 | 827 | |||
GenBank mRNA | AB209261.1 | - | 7 | 836 |