Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THYN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037304.1 | NP_001032381.1 | 3 | 200 | ||
NM_001037305.1 | NP_001032382.1 | 3 | 200 | |||
NM_014174.2 | NP_054893.1 | 2 | 582 | |||
NM_199297.1 | NP_954994.1 | 3 | 272 | |||
NM_199298.1 | NP_954995.1 | 3 | 272 | |||
XM_005271527.3 | XP_005271584.1 | 3 | 235 | |||
XM_005271528.3 | XP_005271585.1 | 3 | 235 | |||
GenBank mRNA | AF059619.1 | - | 3 | 244 | ||
AF087886.1 | - | 3 | 205 | |||
AF161493.1 | - | 2 | 544 | |||
AF182413.1 | - | 3 | 211 | |||
AK308498.1 | - | 2 | 461 | |||
AK311024.1 | - | 2 | 564 | |||
BC006978.1 | - | 3 | 256 | 3682324 | ||
BC093074.1 | - | 3 | 175 | 30717693 | ||
BQ640137.1 | - | 3 | 190 | |||
BQ959408.1 | - | 2 | 585 | |||
BU658940.1 | - | 3 | 261 | |||
BU934600.1 | - | 3 | 202 |