Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014248.3 | NP_055063.1 | 4 | 336 | ||
GenBank mRNA | AF085906.1 | - | 4 | 265 | ||
AF140598.1 | - | 4 | 290 | |||
AF142059.1 | - | 4 | 284 | |||
AK090764.1 | - | 1 | 1654 | |||
AK315722.1 | - | 4 | 295 | |||
AY099360.1 | - | 4 | 247 | |||
BC001466.2 | - | 4 | 289 | 3138751 | ||
BC017370.1 | - | 4 | 306 | |||
CR456560.1 | - | 4 | 302 | |||
DN602016.1 | - | 1 | 14 |