Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYCP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014258.3 | NP_055073.2 | 5 | 425 | ||
XM_011528487.2 | XP_011526789.1 | 5 | 461 | |||
XM_011528488.2 | XP_011526790.1 | 5 | 413 | |||
XM_011528489.2 | XP_011526791.1 | 5 | 456 | |||
XM_011528490.2 | XP_011526792.1 | 5 | 437 | |||
XM_011528493.2 | XP_011526795.1 | 5 | 462 | |||
XM_017027586.1 | XP_016883075.1 | 4 | 657 | |||
XM_017027587.1 | XP_016883076.1 | 4 | 654 | |||
XM_017027588.1 | XP_016883077.1 | 4 | 658 | |||
XM_017027589.1 | XP_016883078.1 | 4 | 658 | |||
XM_017027590.1 | XP_016883079.1 | 5 | 436 | |||
XM_017027592.1 | XP_016883081.1 | 4 | 659 | |||
GenBank mRNA | AK308889.1 | - | 3 | 673 | ||
BC040566.1 | - | 5 | 461 | |||
BC132864.1 | - | 4 | 294 | 40146886 | ||
BC132870.1 | - | 4 | 296 | 40146892 | ||
BC146798.1 | - | 5 | 413 | |||
BX648091.1 | - | 5 | 379 | |||
BX649108.1 | - | 5 | 405 | |||
Y08982.1 | - | 3 | 204 |