Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADAMTS7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014272.4 | NP_055087.2 | 3 | 734 | ||
XM_005254137.3 | XP_005254194.1 | 3 | 735 | |||
GenBank mRNA | AB209497.1 | - | 2 | 388 | ||
AF140675.1 | - | 3 | 503 | |||
AK299844.1 | - | 4 | 669 | |||
AY327122.1 | - | 3 | 501 | |||
BC061631.1 | - | 3 | 714 | 6650221 |