Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAPN6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014289.3 | NP_055104.2 | 4 | 514 | ||
GenBank mRNA | AF029232.1 | - | 4 | 414 | ||
AJ000388.1 | - | 4 | 505 | |||
AK300340.1 | - | 3 | 263 | |||
AK312701.1 | - | 4 | 478 | |||
BC000730.2 | - | 3 | 343 | 3503916 | ||
DA863483.1 | - | 4 | 514 |