Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIK3R5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142633.2 | NP_001136105.1 | 9, 10 | 1023 | ||
NM_001251852.1 | NP_001238781.1 | 8, 9 | 907 | |||
NM_001251853.1 | NP_001238782.1 | 8, 9 | 869 | |||
NM_001251855.1 | NP_001238784.1 | 8, 9 | 848 | |||
NM_014308.3 | NP_055123.2 | 9, 10 | 960 | |||
XM_005256580.3 | XP_005256637.1 | 9, 10 | 1018 | |||
GenBank mRNA | AB028925.1 | - | 8, 9 | 893 | ||
AF128881.1 | - | 9, 10 | 960 | |||
AK309074.1 | - | 8, 9 | 907 | |||
AK314186.1 | - | 9, 10 | 1018 | |||
AY725852.1 | - | 8, 9 | 772 | |||
AY725853.1 | - | 9, 10 | 926 | |||
AY725854.1 | - | 9, 10 | 926 | |||
BC028212.1 | - | 8, 9 | 839 | 5215094 |