Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPL15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014175.3 | NP_054894.1 | 5 | 784 | ||
GenBank mRNA | AF161494.1 | - | 5 | 776 | ||
AK312832.1 | - | 5 | 748 | |||
AL136665.1 | - | 5 | 754 | |||
BC000891.1 | - | 5 | 784 | 3463430 | ||
CR533531.1 | - | 5 | 721 |