Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014252.3 | NP_055067.1 | 6, 7 | 1111 | ||
GenBank mRNA | AF112968.1 | - | 6, 7 | 948 | ||
AF177333.1 | - | 6, 7 | 913 | |||
AK296862.1 | - | 7, 8 | 763 | |||
AK313602.1 | - | 6, 7 | 993 | |||
BC002702.2 | - | 6, 7 | 1025 | 3614008 |