Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TENM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163278.1 | NP_001156750.1 | 1 | 263 | ||
NM_001163279.1 | NP_001156751.1 | 1 | 263 | |||
NM_014253.3 | NP_055068.2 | 1 | 263 | |||
XM_011531230.2 | XP_011529532.1 | 4 | 904 | |||
XM_017029208.1 | XP_016884697.1 | 4 | 904 | |||
XM_017029209.1 | XP_016884698.1 | 5 | 959 | |||
XM_017029210.1 | XP_016884699.1 | 3 | 832 | |||
XM_017029211.1 | XP_016884700.1 | 4 | 910 | |||
XM_017029212.1 | XP_016884701.1 | 4 | 521 | |||
XM_017029213.1 | XP_016884702.1 | 3 | 833 | |||
XM_017029214.1 | XP_016884703.1 | 4 | 904 | |||
XM_017029215.1 | XP_016884704.1 | 4 | 903 | |||
XM_017029216.1 | XP_016884705.1 | 4 | 903 | |||
GenBank mRNA | AF100772.1 | - | 1 | 263 | ||
BC140783.1 | - | 1 | 260 | 9021651 | ||
BC144543.1 | - | 1 | 260 | 9053070 | ||
HE578281.1 | - | 1 | 199 |