Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B3GNT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014256.3 | NP_055071.2 | 3 | 1394 | ||
XM_011527626.2 | XP_011525928.1 | 3 | 1353 | |||
GenBank mRNA | AB015630.1 | - | 3 | 1364 | ||
AB049585.1 | - | 3 | 1349 | |||
AK075434.1 | - | 4 | 944 | |||
AK225421.1 | - | 3 | 1361 | |||
AY358955.1 | - | 3 | 1349 |