Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLEC4M | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144904.1 | NP_001138376.1 | 7 | 1678 | ||
NM_001144905.1 | NP_001138377.1 | 8 | 1759 | |||
NM_001144906.1 | NP_001138378.1 | 6 | 1423 | |||
NM_001144907.1 | NP_001138379.1 | 7 | 1630 | |||
NM_001144908.1 | NP_001138380.1 | 6 | 1442 | |||
NM_001144909.1 | NP_001138381.1 | 8 | 1693 | |||
NM_001144910.1 | NP_001138382.1 | 8 | 1762 | |||
NM_001144911.1 | NP_001138383.1 | 5 | 1634 | |||
NM_014257.4 | NP_055072.3 | 7 | 1831 | |||
NR_026707.1 | 6 | 2279 | ||||
NR_026708.1 | 5 | 2166 | ||||
NR_026709.1 | 4 | 2103 | ||||
XM_006722611.2 | XP_006722674.1 | 7 | 1763 | |||
XM_006722612.3 | XP_006722675.1 | 6 | 1837 | |||
XM_006722613.3 | XP_006722676.1 | 6 | 1839 | |||
XM_006722614.3 | XP_006722677.1 | 5 | 1755 | |||
XM_006722615.2 | XP_006722678.1 | 6 | 1718 | |||
GenBank mRNA | AB015629.1 | - | 5 | 1540 | ||
AF245219.2 | - | 8 | 1407 | |||
AF290887.1 | - | 8 | 1697 | |||
BC038851.1 | - | 7 | 1785 | 5166966 | ||
BX102225.1 | - | 1 | 351 |