Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYCP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014258.3 | NP_055073.2 | 24 | 2238 | ||
XM_011528487.2 | XP_011526789.1 | 24 | 2274 | |||
XM_011528488.2 | XP_011526790.1 | 24 | 2226 | |||
XM_011528489.2 | XP_011526791.1 | 24 | 2269 | |||
XM_011528490.2 | XP_011526792.1 | 24 | 2250 | |||
XM_011528493.2 | XP_011526795.1 | 23 | 2189 | |||
XM_017027586.1 | XP_016883075.1 | 23 | 2470 | |||
XM_017027587.1 | XP_016883076.1 | 23 | 2467 | |||
XM_017027588.1 | XP_016883077.1 | 23 | 2471 | |||
XM_017027589.1 | XP_016883078.1 | 23 | 2471 | |||
XM_017027590.1 | XP_016883079.1 | 23 | 2163 | |||
XM_017027591.1 | XP_016883080.1 | 15 | 1569 | |||
XM_017027592.1 | XP_016883081.1 | 23 | 2472 | |||
GenBank mRNA | AL080226.1 | - | 12 | 1172 | ||
BC040566.1 | - | 24 | 2274 | |||
BC132864.1 | - | 23 | 2107 | 40146886 | ||
BC132870.1 | - | 23 | 2109 | 40146892 | ||
BC146798.1 | - | 24 | 2226 | |||
BX648091.1 | - | 24 | 2192 | |||
BX649108.1 | - | 24 | 2218 | |||
Y08982.1 | - | 22 | 2017 |