Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADAMTS7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014272.4 | NP_055087.2 | 9, 10 | 1709 | ||
XM_005254137.3 | XP_005254194.1 | 9, 10 | 1710 | |||
GenBank mRNA | AF140675.1 | - | 9, 10 | 1478 | ||
AY327122.1 | - | 9, 10 | 1476 | |||
BC061631.1 | - | 9, 10 | 1689 | 6650221 |
Gene Symbol: ADAMTS7P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045529.3 | 9, 10 | 1221 |