Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSPA4L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317381.1 | NP_001304310.1 | 12 | 1651 | ||
NM_001317382.1 | NP_001304311.1 | 11 | 1370 | |||
NM_001317383.1 | NP_001304312.1 | 10 | 1538 | |||
NM_014278.3 | NP_055093.2 | 11 | 1661 | |||
XM_011531745.2 | XP_011530047.1 | 11 | 1730 | |||
GenBank mRNA | AB023421.1 | - | 11 | 1373 | ||
AK027822.1 | - | 11 | 1548 | |||
AK302115.1 | - | 11 | 1344 | |||
AK303053.1 | - | 12 | 1651 | |||
AK307464.1 | - | 10 | 1425 | |||
AY290862.1 | - | 11 | 1527 | |||
BC040560.1 | - | 11 | 1572 | 5270824 | ||
EF197155.1 | - | 11 | 1319 |