Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NCDN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014839.1 | NP_001014839.1 | 8 | 3261 | ||
NM_001014841.1 | NP_001014841.1 | 7 | 3193 | |||
NM_014284.2 | NP_055099.1 | 7 | 3533 | |||
GenBank mRNA | AB011179.1 | - | 7 | 3140 | ||
BC024592.1 | - | 7 | 3507 | 3914058 |