Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRAM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317804.1 | NP_001304733.1 | 11 | 1212 | ||
NM_001317805.1 | NP_001304734.1 | 10 | 1727 | |||
NM_014294.5 | NP_055109.1 | 10 | 1295 | |||
XM_017013266.1 | XP_016868755.1 | 9 | 1167 | |||
XM_017013267.1 | XP_016868756.1 | 9 | 1511 | |||
GenBank mRNA | AK289397.1 | - | 10 | 1155 | ||
AK292777.1 | - | 10 | 1079 | |||
AK303411.1 | - | 11 | 1140 | |||
BC000687.2 | - | 10 | 1117 | 3347823 | ||
BC020283.1 | - | 10 | 1110 | |||
BC037738.1 | - | 10 | 1207 | 5261490 | ||
BT007359.1 | - | 10 | 1035 | |||
CR541654.1 | - | 10 | 1035 | |||
CR541739.1 | - | 10 | 1035 | |||
X63679.1 | - | 10 | 1156 |