Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM50A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014313.3 | NP_055128.1 | 7 | 2077 | ||
XM_005245817.1 | XP_005245874.1 | 6 | 1948 | |||
XM_011541159.1 | XP_011539461.1 | 7 | 2271 | |||
GenBank mRNA | AF081282.1 | - | 7 | 2005 | ||
AL136627.1 | - | 7 | 2059 | |||
AY071927.1 | - | 7 | 2054 | |||
BC066908.1 | - | 7 | 2016 | 4817784 | ||
BX537683.1 | - | 1 | 5001 | |||
JX644905.1 | - | 7 | 2077 | |||
JX644906.1 | - | 7 | 2077 |