Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ORC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014321.3 | NP_055136.1 | 7 | 758 | ||
NR_037620.1 | 7 | 817 | ||||
GenBank mRNA | AF139658.1 | - | 7 | 746 | ||
AK021946.1 | - | 7 | 719 | |||
AK024019.1 | - | 7 | 752 | |||
BC039032.1 | - | 7 | 738 | 5577396 | ||
BC063565.1 | - | 7 | 727 | 3460593 |