Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMPX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014332.2 | NP_055147.1 | 4 | 395 | ||
NR_045617.1 | 4 | 395 | ||||
GenBank mRNA | AF129505.1 | - | 4 | 347 | ||
AJ250584.1 | - | 4 | 347 | |||
AK312134.1 | - | 4 | 395 | |||
BC005948.1 | - | 4 | 276 | 4246501 | ||
BF126275.1 | - | 4 | 257 |