Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PISD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001326413.1 | NP_001313342.1 | 3, 4 | 770 | ||
NM_001326414.1 | NP_001313343.1 | 4, 5 | 850 | |||
NM_001326415.1 | NP_001313344.1 | 4, 5 | 1024 | |||
NM_001326416.1 | NP_001313345.1 | 6, 7 | 1213 | |||
NM_001326417.1 | NP_001313346.1 | 5, 6 | 1104 | |||
NM_001326419.1 | NP_001313348.1 | 5, 6 | 1104 | |||
NM_014338.3 | NP_055153.1 | 4, 5 | 656 | |||
NM_178022.1 | NP_821141.1 | 3, 4 | 565 | |||
GenBank mRNA | AK298474.1 | - | 3, 4 | 565 | ||
BC001482.2 | - | 4, 5 | 656 | 3508281 |